accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 83905-98-0

IUPAC Name: (11-hydroxyundecyl)phosphonic acid
Synonyms: 11-hydroxyundecylphosphonic acid
Molecular Weight (g/mol): 252.29
Molecular Formula: C11H25O4P
InChi Key: PPCDEFQVKBXBPS-UHFFFAOYSA-N
SMILES: OCCCCCCCCCCCP(O)(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results