CAS RN 84-17-3
IUPAC Name:
4-[(2E,4E)-4-(4-hydroxyphenyl)hexa-2,4-dien-3-yl]phenol
Synonyms:
dienestrol dienestrol, (Z,E)- para-dien dinestrol gynefollin sexadien (Z,Z)-isomer dienestrol dienoestrolum dienestrol, (E,E)- retalon
Molecular Weight (g/mol):
266.34
Molecular Formula:
C18H18O2
InChi Key:
NFDFQCUYFHCNBW-SCGPFSFSSA-N
SMILES:
C\C=C(\C(=C\C)\C1=CC=C(O)C=C1)/C1=CC=C(O)C=C1
1
–
1
of
1
results
1
Dienestrol 96.0+%, TCI America™
CAS: 84-17-3 Molecular Formula: C18H18O2 Molecular Weight (g/mol): 266.34 MDL Number: MFCD00050983 InChI Key: NFDFQCUYFHCNBW-SCGPFSFSSA-N PubChem CID: 667476 IUPAC Name: 4-[(2E,4E)-4-(4-hydroxyphenyl)hexa-2,4-dien-3-yl]phenol SMILES: CC=C(C1=CC=C(C=C1)O)C(=CC)C2=CC=C(C=C2)O