accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3OCH3OCH3O(Z)(Z)

CAS RN 84-19-5

IUPAC Name: 4-[(2Z,4Z)-4-[4-(acetyloxy)phenyl]hexa-2,4-dien-3-yl]phenyl acetate
Synonyms: dienestrol di(acetate)
Molecular Weight (g/mol): 350.41
Molecular Formula: C22H22O4
InChi Key: YWLLGDVBTLPARJ-CMDWGMEOSA-N
SMILES: C\C=C(/C(=C\C)/C1=CC=C(OC(C)=O)C=C1)\C1=CC=C(OC(C)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results