CAS RN 84-59-3
IUPAC Name:
2,6-dibromonaphthalene-1,5-diol
Synonyms:
2,6-dibromonaphthalene-1,5-diol
Molecular Weight (g/mol):
317.96
Molecular Formula:
C10H6Br2O2
InChi Key:
GHJUWGHWJYULLK-UHFFFAOYSA-N
SMILES:
OC1=C2C=CC(Br)=C(O)C2=CC=C1Br
1
–
1
of
1
results
1
2,6-Dibromo-1,5-dihydroxynaphthalene 93.0+%, TCI America™
CAS: 84-59-3 Molecular Formula: C10H6Br2O2 Molecular Weight (g/mol): 317.964 MDL Number: MFCD00021473 InChI Key: GHJUWGHWJYULLK-UHFFFAOYSA-N Synonym: 2,6-Dibromo-1,5-naphthalenediol PubChem CID: 96260 IUPAC Name: 2,6-dibromonaphthalene-1,5-diol SMILES: C1=CC(=C(C2=C1C(=C(C=C2)Br)O)O)Br