accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84-61-7

IUPAC Name: 1,2-dicyclohexyl benzene-1,2-dicarboxylate
Synonyms: dicyclohexyl phthalate unimoll 66 ergoplast fdc ergoplast.fdc howflex CP
Molecular Weight (g/mol): 330.42
Molecular Formula: C20H26O4
InChi Key: VOWAEIGWURALJQ-UHFFFAOYSA-N
SMILES: O=C(OC1CCCCC1)C1=CC=CC=C1C(=O)OC1CCCCC1
Molecular Weight (g/mol) 
  • (6)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (4)
  • (2)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (6)
  • (4)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (6)