accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)OOHNCH3OO

CAS RN 84000-11-3

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-3-methylbutanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}-3-methylbutanoic acid| FMOC-N-ME-VAL-OH
Molecular Weight (g/mol): 353.42
Molecular Formula: C21H23NO4
InChi Key: YCXXXPZNQXXRIG-IBGZPJMESA-N
SMILES: CC(C)[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Quantity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)