accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNHNH

CAS RN 841-77-0

IUPAC Name: 1-(diphenylmethyl)piperazin-1-ium chloride
Synonyms: diphenylmethylpiperazine
Molecular Weight (g/mol): 288.82
Molecular Formula: C17H21ClN2
InChi Key: DLCXUFKAJDKSDD-UHFFFAOYSA-N
SMILES: [Cl-].C1C[NH+](CCN1)C(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)