CAS RN 84102-58-9
CAS RN 84102-58-9
IUPAC Name:
2-(4-aminophenyl)-1-benzofuran-5-amine
Synonyms:
2-(4-aminophenyl)-1-benzofuran-5-amine
Molecular Weight (g/mol):
224.26
Molecular Formula:
C14H12N2O
InChi Key:
FKMCNLLISWDXJQ-UHFFFAOYSA-N
SMILES:
NC1=CC=C(C=C1)C1=CC2=CC(N)=CC=C2O1
1
–
1
of
1
results
1
5-Amino-2-(4-aminophenyl)benzofuran 98.0+%, TCI America™
CAS: 84102-58-9 Molecular Formula: C14H12N2O Molecular Weight (g/mol): 224.26 MDL Number: MFCD10000946 InChI Key: FKMCNLLISWDXJQ-UHFFFAOYSA-N Synonym: 2-(4-Aminophenyl)-5-benzofuranamine PubChem CID: 11106990 IUPAC Name: 2-(4-aminophenyl)-1-benzofuran-5-amine SMILES: NC1=CC=C(C=C1)C1=CC2=CC(N)=CC=C2O1