accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84163-16-6

IUPAC Name: 5-fluoro-3-(piperidin-4-yl)-1,2-benzoxazole hydrochloride
Synonyms: 5-fluoro-3-(piperidin-4-yl)-1,2-benzoxazole hydrochloride
Molecular Weight (g/mol): 256.71
Molecular Formula: C12H14ClFN2O
InChi Key: HSOJFILHZZEWJT-UHFFFAOYSA-N
SMILES: Cl.FC1=CC=C2ON=C(C3CCNCC3)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results