accessibility menu, dialog, popup

UserName

Viewing profile as user:
FONHN

CAS RN 84163-77-9

IUPAC Name: 6-fluoro-3-(piperidin-4-yl)-1,2-benzoxazole
Synonyms: 6-fluoro-3-(piperidin-4-yl)-1,2-benzoxazole
Molecular Weight (g/mol): 220.25
Molecular Formula: C12H13FN2O
InChi Key: MRMGJMGHPJZSAE-UHFFFAOYSA-N
SMILES: FC1=CC=C2C(ON=C2C2CCNCC2)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results