accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOHOO

CAS RN 84182-46-7

IUPAC Name: 3-(benzyloxy)cyclobutane-1,1-dicarboxylic acid
Synonyms: 3-(benzyloxy)cyclobutane-1,1-dicarboxylic acid
Molecular Weight (g/mol): 250.25
Molecular Formula: C13H14O5
InChi Key: LYRCCQROQBVCRV-UHFFFAOYSA-N
SMILES: OC(=O)C1(CC(C1)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results