accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 842-15-9

IUPAC Name: potassium 7-amino-3-sulfonaphthalene-1-sulfonate
Synonyms: potassium 7-amino-3-sulfonaphthalene-1-sulfonate
Molecular Weight (g/mol): 341.39
Molecular Formula: C10H8KNO6S2
InChi Key: GOPRYBNRWIBTIH-UHFFFAOYSA-M
SMILES: [K+].NC1=CC=C2C=C(C=C(C2=C1)S([O-])(=O)=O)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results