accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOOOOOOHOHOOOHOHOOOHOOHOOHOOHOOOHOHOOOOHOHHOHOHOOHOHOHHOOH

CAS RN 84216-71-7

IUPAC Name: 36,38,39,40,41,42,43,44,45,46,47,48,49-tridecahydroxy-5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontan-37-yl 4-methylbenzene-1-sulfonate
Synonyms: 36,38,39,40,41,42,43,44,45,46,47,48,49-tridecahydroxy-5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontan-37-yl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 1289.17
Molecular Formula: C49H76O37S
InChi Key: VGZWDYWXWGKKEE-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC1C(O)C2OC3OC(CO)C(OC4OC(CO)C(OC5OC(CO)C(OC6OC(CO)C(OC7OC(CO)C(OC8OC(CO)C(OC1OC2CO)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results