accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOONHSOOFFFFFF

CAS RN 84246-29-7

IUPAC Name: 4,4,5,5,6,6-hexafluoro-1λ⁶,3λ⁶,2-dithiazinane-1,1,3,3-tetrone
Synonyms: 4,4,5,5,6,6-hexafluoro-1λ⁶,3λ⁶,2-dithiazinane-1,1,3,3-tetrone
Molecular Weight (g/mol): 293.15
Molecular Formula: C3HF6NO4S2
InChi Key: WOAGDWWRYOZHDS-UHFFFAOYSA-N
SMILES: FC1(F)C(F)(F)S(=O)(=O)NS(=O)(=O)C1(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results