accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2BrN

CAS RN 84249-14-9

IUPAC Name: 4-bromopyridin-2-amine
Synonyms: 4-bromopyridin-2-amine
Molecular Weight (g/mol): 173.01
Molecular Formula: C5H5BrN2
InChi Key: BAQKUNMKVAPWGU-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=CC=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)