accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 842974-69-0

IUPAC Name: 2-(morpholin-4-yl)pyrimidine-5-carbaldehyde
Synonyms: 2-(morpholin-4-yl)pyrimidine-5-carbaldehyde
Molecular Weight (g/mol): 193.21
Molecular Formula: C9H11N3O2
InChi Key: XDMVIYFKCVJZHK-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(N=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results