accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COOHNCH3OHOHO

CAS RN 84311-18-2

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-hydroxy-2-methylpropanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-hydroxy-2-methylpropanoic acid
Molecular Weight (g/mol): 219.24
Molecular Formula: C9H17NO5
InChi Key: FWRXDSRYWWYTPD-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(C)(CO)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results