accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84315-23-1

IUPAC Name: (2E)-3-(3,5-difluorophenyl)prop-2-enoate
Synonyms: (2E)-3-(3,5-difluorophenyl)prop-2-enoate
Molecular Weight (g/mol): 183.13
Molecular Formula: C9H5F2O2
InChi Key: MBAWRXICVNIUGY-OWOJBTEDSA-M
SMILES: [O-]C(=O)\C=C\C1=CC(F)=CC(F)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results