accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFBrONa

CAS RN 84349-27-9

IUPAC Name: sodium 2-bromo-2,2-difluoroacetate
Synonyms: sodium 2-bromo-2,2-difluoroacetate
Molecular Weight (g/mol): 196.91
Molecular Formula: C2BrF2NaO2
InChi Key: CAQKQIYWKXZJGD-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)C(F)(F)Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results