accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84387-61-1

IUPAC Name: 1-(chloromethyl)-1H-1,2,4-triazole hydrochloride
Synonyms: 1-(chloromethyl)-1,2,4-triazole hydrochloride
Molecular Weight (g/mol): 153.99
Molecular Formula: C3H5Cl2N3
InChi Key: GXVWDLWTFIDZLR-UHFFFAOYSA-N
SMILES: Cl.ClCN1C=NC=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results