accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNSNOO

CAS RN 84387-89-3

IUPAC Name: 5-nitro-1,2-benzothiazol-3-amine
Synonyms: 5-nitro-1,2-benzothiazol-3-amine
Molecular Weight (g/mol): 195.2
Molecular Formula: C7H5N3O2S
InChi Key: LDTCWISGJYTXDC-UHFFFAOYSA-N
SMILES: NC1=NSC2=CC=C(C=C12)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results