accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84405-44-7

IUPAC Name: 2,7-dibromo-9,10-dihydrophenanthrene-9,10-dione
Synonyms: 2,7-dibromophenanthrene-9,10-dione
Molecular Weight (g/mol): 366.01
Molecular Formula: C14H6Br2O2
InChi Key: LTZIIBSPYWTOQV-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C3=CC=C(Br)C=C3C(=O)C(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results