accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOOH2NBr

CAS RN 84487-04-7

IUPAC Name: 6-bromo-3-nitropyridin-2-amine
Synonyms: 6-bromo-3-nitropyridin-2-amine
Molecular Weight (g/mol): 218.01
Molecular Formula: C5H4BrN3O2
InChi Key: YRXZIHANXVKUHP-UHFFFAOYSA-N
SMILES: NC1=C(C=CC(Br)=N1)[N+]([O-])=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results