accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 844891-01-6

IUPAC Name: 4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)isoquinoline
Synonyms: 4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)isoquinoline
Molecular Weight (g/mol): 241.1
Molecular Formula: C14H16BNO2
InChi Key: AQEZCUCPPAKCLQ-UHFFFAOYSA-N
SMILES: CC1(C)COB(OC1)C1=CN=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results