accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 844891-03-8

IUPAC Name: 1,3-dimethyl-1H-pyrazole-5-carbothioamide
Synonyms: 2,5-dimethylpyrazole-3-carbothioamide
Molecular Weight (g/mol): 155.22
Molecular Formula: C6H9N3S
InChi Key: RVWYPAZKPYVUDC-UHFFFAOYSA-N
SMILES: CN1N=C(C)C=C1C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results