accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 844891-09-4

IUPAC Name: tert-butyl 4-[(4-formylphenyl)methyl]piperazine-1-carboxylate
Synonyms: tert-butyl 4-[(4-formylphenyl)methyl]piperazine-1-carboxylate
Molecular Weight (g/mol): 304.39
Molecular Formula: C17H24N2O3
InChi Key: UMXKYTSTYWRBBK-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC2=CC=C(C=O)C=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results