accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84494-72-4

IUPAC Name: ethyl (4E,7E,10E,13E,16E,19E)-docosa-4,7,10,13,16,19-hexaenoate
Synonyms: ethyl (4E,7E,10E,13E,16E,19E)-docosa-4,7,10,13,16,19-hexaenoate
Molecular Weight (g/mol): 356.55
Molecular Formula: C24H36O2
InChi Key: ITNKVODZACVXDS-YATCGRJWSA-N
SMILES: CCOC(=O)CC\C=C\C\C=C\C\C=C\C\C=C\C\C=C\C\C=C\CC
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results