CAS RN 84508-45-2
IUPAC Name:
ethyl acetate
Synonyms:
ethyl acetate caswell no. 429 acetato de etilo ethyl acetate (natural) acetate d'ethyle etile (acetato di) acetidin acetic ester essigester ethyl acetate (substance)
Molecular Weight (g/mol):
88.11
Molecular Formula:
C4H8O2
InChi Key:
XEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILES:
CCOC(C)=O
1
–
1
of
1
results
1
Ethyl Acetate-1,2-C2, SPEX CertiPrep™
CAS: 84508-45-2 Molecular Formula: C4H8O2 Molecular Weight (g/mol): 90.091 InChI Key: XEKOWRVHYACXOJ-NDLBAUGKSA-N PubChem CID: 12206864 IUPAC Name: ethyl acetate SMILES: CCOC(=O)C