CAS RN 845714-00-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Cdc7/Cdk9 Inhibitor
The Cdc7/Cdk9 Inhibitor, also referenced under CAS 845714-00-3, controls the biological activity of Cdc7/Cdk9. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.