accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrFF

CAS RN 845827-13-6

IUPAC Name: 5-bromo-2-(difluoromethyl)pyridine
Synonyms: 5-bromo-2-(difluoromethyl)pyridine
Molecular Weight (g/mol): 208.01
Molecular Formula: C6H4BrF2N
InChi Key: QXLZRIGSWWQOLG-UHFFFAOYSA-N
SMILES: FC(F)C1=NC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results