accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 845870-55-5

IUPAC Name: N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 233.12
Molecular Formula: C13H20BNO2
InChi Key: WWGNYCWKFZIQQS-UHFFFAOYSA-N
SMILES: CNC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results