accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNFFFOBr

CAS RN 845885-81-6

IUPAC Name: 2-bromo-1-{4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}ethan-1-one
Synonyms: 2-bromo-1-{4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}ethanone
Molecular Weight (g/mol): 364.18
Molecular Formula: C13H9BrF3NOS
InChi Key: XGHUMHHAMHEPCS-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C1=CC=C(C=C1)C(F)(F)F)C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results