accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(R)OOHNHOO

CAS RN 84624-17-9

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylbutanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-methylbutanoic acid|FMOC-D-VAL-OH
Molecular Weight (g/mol): 339.39
Molecular Formula: C20H21NO4
InChi Key: UGNIYGNGCNXHTR-GOSISDBHSA-N
SMILES: CC(C)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Quantity 
  • (2)
  • (3)
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)