accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHN(S)OHONH2

CAS RN 84624-28-2

IUPAC Name: (2S)-2-amino-6-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: (2S)-2-amino-6-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 368.43
Molecular Formula: C21H24N2O4
InChi Key: RAQBUPMYCNRBCQ-TWYLJJHKNA-N
SMILES: N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)