CAS RN 84635-54-1
IUPAC Name:
4,5-bis(acetyloxy)-2-methyl-6-(methylsulfanyl)oxan-3-yl acetate
Synonyms:
4,5-bis(acetyloxy)-2-methyl-6-(methylsulfanyl)oxan-3-yl acetate
Molecular Weight (g/mol):
320.36
Molecular Formula:
C13H20O7S
InChi Key:
JOLBZRHXWOHZNS-UHFFFAOYNA-N
SMILES:
CSC1OC(C)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
1
–
1
of
1
results
1
Methyl 2,3,4-Tri-O-acetyl-1-thio-beta-L-fucopyranoside 98.0+%, TCI America™
CAS: 84635-54-1 Molecular Formula: C13H20O7S Molecular Weight (g/mol): 320.356 MDL Number: MFCD00080803 InChI Key: JOLBZRHXWOHZNS-MCNNAKBESA-N PubChem CID: 14131721 IUPAC Name: [(2S,3R,4R,5S,6R)-4,5-diacetyloxy-2-methyl-6-methylsulfanyloxan-3-yl] acetate SMILES: CC1C(C(C(C(O1)SC)OC(=O)C)OC(=O)C)OC(=O)C