accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNHOO

CAS RN 84677-06-5

IUPAC Name: 1-{[(benzyloxy)carbonyl]amino}cyclopropane-1-carboxylic acid
Synonyms: 1-{[(benzyloxy)carbonyl]amino}cyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 235.24
Molecular Formula: C12H13NO4
InChi Key: KHINKCGJKZSHAJ-UHFFFAOYSA-N
SMILES: OC(=O)C1(CC1)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)