accessibility menu, dialog, popup

UserName

Viewing profile as user:
FePPt-Butylt-Butylt-Butylt-Butyl

CAS RN 84680-95-5

IUPAC Name: 1,1'-Bis(di-tert-butylphosphino)ferrocene
Synonyms: —
Molecular Weight (g/mol): 474.43
Molecular Formula: C26H44FeP2
InChi Key: —
SMILES: [Fe].CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)