accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 847-83-6

IUPAC Name: (4-methoxyphenyl)diphenylmethanol
Synonyms: (4-methoxyphenyl)diphenylmethanol
Molecular Weight (g/mol): 290.36
Molecular Formula: C20H18O2
InChi Key: WCRRRAKYYPJJMP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)