accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHO

CAS RN 847417-37-2

IUPAC Name: tert-butyl N-[4-(2-oxoethyl)cyclohexyl]carbamate
Synonyms: tert-butyl N-[4-(2-oxoethyl)cyclohexyl]carbamate
Molecular Weight (g/mol): 241.33
Molecular Formula: C13H23NO3
InChi Key: OPGBSEXLSWYFOR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1CCC(CC=O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)