accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FFBr

CAS RN 847502-81-2

IUPAC Name: 1-bromo-3,4-difluoro-2-methylbenzene
Synonyms: 1-bromo-3,4-difluoro-2-methylbenzene
Molecular Weight (g/mol): 207.02
Molecular Formula: C7H5BrF2
InChi Key: YXTCBTSPHSNDHD-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=CC(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results