accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NNOOONi(E/Z)(E/Z)

CAS RN 847654-16-4

IUPAC Name: nickel(2+) 2-{[(2-{[2-(dibutylamino)-1-oxidoethylidene]amino}phenyl)(phenyl)methylidene]amino}acetate
Synonyms: nickel(2+) 2-{[(2-{[2-(dibutylamino)-1-oxidoethylidene]amino}phenyl)(phenyl)methylidene]amino}acetate
Molecular Weight (g/mol): 480.23
Molecular Formula: C25H31N3NiO3
InChi Key: KZZGXIZCIUYKSB-UHFFFAOYSA-L
SMILES: [Ni++].CCCCN(CCCC)CC([O-])=NC1=CC=CC=C1C(=NCC([O-])=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results