accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONS

CAS RN 847956-27-8

IUPAC Name: 3-(1,3-thiazol-2-yl)benzoic acid
Synonyms: 3-(1,3-thiazol-2-yl)benzoic acid
Molecular Weight (g/mol): 205.23
Molecular Formula: C10H7NO2S
InChi Key: BLFSSABBFANDKK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results