accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 848366-28-9

IUPAC Name: 5-bromo-3-chloro-2-methoxypyridine
Synonyms: 5-bromo-3-chloro-2-methoxypyridine
Molecular Weight (g/mol): 222.47
Molecular Formula: C6H5BrClNO
InChi Key: XCHXNAQRMMXBGN-UHFFFAOYSA-N
SMILES: COC1=NC=C(Br)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results