accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84852-53-9

IUPAC Name: 1,2,3,4,5-pentabromo-6-[2-(2,3,4,5,6-pentabromophenyl)ethyl]benzene
Synonyms: 1,2,3,4,5-pentabromo-6-[2-(2,3,4,5,6-pentabromophenyl)ethyl]benzene
Molecular Weight (g/mol): 971.23
Molecular Formula: C14H4Br10
InChi Key: BZQKBFHEWDPQHD-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C(Br)=C(CCC2=C(Br)C(Br)=C(Br)C(Br)=C2Br)C(Br)=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results