accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNH3CCH3OPOOOH3C

CAS RN 848641-69-0

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium diethyl phosphate
Synonyms: 1-ethyl-3-methylimidazolium diethyl phosphate
Molecular Weight (g/mol): 264.26
Molecular Formula: C10H21N2O4P
InChi Key: HQWOEDCLDNFWEV-UHFFFAOYSA-M
SMILES: CCN1C=C[N+](C)=C1.CCOP([O-])(=O)OCC
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)