accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOH3CHOOH

CAS RN 849062-20-0

IUPAC Name: {4'-propoxy-[1,1'-biphenyl]-4-yl}boronic acid
Synonyms: 4'-propoxy-[1,1'-biphenyl]-4-ylboronic acid
Molecular Weight (g/mol): 256.11
Molecular Formula: C15H17BO3
InChi Key: ZJCHLFOLMBDUPR-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C(C=C1)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results