accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClClOHN

CAS RN 849107-20-6

IUPAC Name: hydrogen 4-(2-chlorophenoxy)piperidine chloride
Synonyms: hydrogen 4-(2-chlorophenoxy)piperidine chloride
Molecular Weight (g/mol): 248.15
Molecular Formula: C11H15Cl2NO
InChi Key: OWOPJLHKPATCOS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClC1=CC=CC=C1OC1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results