accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 84964-24-9

IUPAC Name: 1-(3-bromophenyl)piperidine
Synonyms: 1-(3-bromophenyl)piperidine
Molecular Weight (g/mol): 240.14
Molecular Formula: C11H14BrN
InChi Key: PMTXPOLLPCSCBE-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results