accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNOHOH2O

CAS RN 849776-47-2

IUPAC Name: 1H-1,3-benzodiazole-2-carboxylic acid hydrate
Synonyms: 1H-1,3-benzodiazole-2-carboxylic acid hydrate
Molecular Weight (g/mol): 180.16
Molecular Formula: C8H8N2O3
InChi Key: YQHUUYLPOCNBKD-UHFFFAOYSA-N
SMILES: O.OC(=O)C1=NC2=CC=CC=C2N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results