accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 849776-88-1

IUPAC Name: 5-methyl-1-phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 5-methyl-1-phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol): 284.17
Molecular Formula: C16H21BN2O2
InChi Key: XSUARWLKVRBBKT-UHFFFAOYSA-N
SMILES: CC1=C(C=NN1C1=CC=CC=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results